Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCCTGTAAGACAAATGGGGTAACA[C/T]AGATCTTTTCTCTATTTTCCAAATA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164720 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ETS1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ETS1 - ETS proto-oncogene 1, transcription factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143820.1 | Intron | NP_001137292.1 | ||||
NM_001162422.1 | Intron | NP_001155894.1 | ||||
NM_005238.3 | Intron | NP_005229.1 | ||||
XM_011542649.2 | Intron | XP_011540951.1 | ||||
XM_011542650.1 | Intron | XP_011540952.1 | ||||
XM_011542651.1 | Intron | XP_011540953.1 | ||||
XM_011542652.1 | Intron | XP_011540954.1 | ||||
XM_017017314.1 | Intron | XP_016872803.1 | ||||
XM_017017315.1 | Intron | XP_016872804.1 | ||||
XM_017017316.1 | Intron | XP_016872805.1 | ||||
XM_017017317.1 | Intron | XP_016872806.1 |
Set Membership: |
HapMap |