Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609023 MIM: 610364 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR6513 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MIR6513 - microRNA 6513 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PNKD - paroxysmal nonkinesigenic dyskinesia | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077399.2 | Intron | NP_001070867.1 | ||||
NM_015488.4 | Intron | NP_056303.3 | ||||
NM_022572.4 | Intron | NP_072094.1 | ||||
XM_017003771.1 | Intron | XP_016859260.1 | ||||
XM_017003772.1 | Intron | XP_016859261.1 |
TMBIM1 - transmembrane BAX inhibitor motif containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321427.1 | Intron | NP_001308356.1 | ||||
NM_001321428.1 | Intron | NP_001308357.1 | ||||
NM_001321429.1 | Intron | NP_001308358.1 | ||||
NM_001321430.1 | Intron | NP_001308359.1 | ||||
NM_001321432.1 | Intron | NP_001308361.1 | ||||
NM_001321433.1 | Intron | NP_001308362.1 | ||||
NM_001321435.1 | Intron | NP_001308364.1 | ||||
NM_001321436.1 | Intron | NP_001308365.1 | ||||
NM_001321438.1 | Intron | NP_001308367.1 | ||||
NM_022152.5 | Intron | NP_071435.2 | ||||
XM_011511627.1 | Intron | XP_011509929.1 |