Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAATCCCTCAACTACGGCTGCATCG[C/T]TGAGAATCCACAGACACACGAGGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615495 MIM: 615950 | ||||||||||||||||||||
Literature Links: |
GMPPA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GMPPA - GDP-mannose pyrophosphorylase A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013335.3 | 490 | Missense Mutation | GCT,GTT | A,V 156 | NP_037467.2 | |
NM_205847.2 | 490 | Missense Mutation | GCT,GTT | A,V 156 | NP_995319.1 | |
XM_005246483.3 | 490 | Missense Mutation | GCT,GTT | A,V 156 | XP_005246540.1 | |
XM_005246486.3 | 490 | Missense Mutation | GCT,GTT | A,V 156 | XP_005246543.1 | |
XM_011511032.2 | 490 | Missense Mutation | GCT,GTT | A,V 156 | XP_011509334.1 | |
XM_017003906.1 | 490 | Missense Mutation | GCT,GTT | A,V 156 | XP_016859395.1 | |
XM_017003907.1 | 490 | Missense Mutation | GCT,GTT | A,V 156 | XP_016859396.1 | |
XM_017003908.1 | 490 | Missense Mutation | GCT,GTT | A,V 156 | XP_016859397.1 | |
XM_017003909.1 | 490 | Missense Mutation | GCT,GTT | A,V 156 | XP_016859398.1 | |
XM_017003910.1 | 490 | Missense Mutation | GCT,GTT | A,V 156 | XP_016859399.1 | |
XM_017003911.1 | 490 | Missense Mutation | GCT,GTT | A,V 156 | XP_016859400.1 | |
XM_017003912.1 | 490 | Missense Mutation | GCT,GTT | A,V 156 | XP_016859401.1 | |
XM_017003913.1 | 490 | Missense Mutation | GCT,GTT | A,V 156 | XP_016859402.1 | |
XM_017003914.1 | 490 | Missense Mutation | GCT,GTT | A,V 31 | XP_016859403.1 | |
XM_017003915.1 | 490 | Missense Mutation | GCT,GTT | A,V 31 | XP_016859404.1 | |
XM_017003916.1 | 490 | Missense Mutation | GCT,GTT | A,V 31 | XP_016859405.1 | |
XM_017003917.1 | 490 | Missense Mutation | GCT,GTT | A,V 31 | XP_016859406.1 |
LOC100996693 - CAVP-target protein-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPEG - SPEG complex locus | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |