Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGCCTCTTAACAAATAGGGTCTTC[G/T]AGGATGCTGACATAGGATTAGATGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615478 | ||||||||||||||||||||
Literature Links: |
NYAP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NYAP2 - neuronal tyrosine-phosphorylated phosphoinositide-3-kinase adaptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020864.1 | Intron | NP_065915.1 | ||||
XM_005246708.3 | Intron | XP_005246765.1 | ||||
XM_011511524.1 | Intron | XP_011509826.1 | ||||
XM_017004564.1 | Intron | XP_016860053.1 |