Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGACGCAGCGGCTTCGGACCCTGC[C/T]CCGGGAAGTCAGCCGTGAGACCGGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
20 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605078 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TARDBP PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TARDBP - TAR DNA binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007375.3 | Intron | NP_031401.1 | ||||
XM_017000863.1 | Intron | XP_016856352.1 | ||||
XM_017000864.1 | Intron | XP_016856353.1 | ||||
XM_017000865.1 | Intron | XP_016856354.1 | ||||
XM_017000866.1 | Intron | XP_016856355.1 | ||||
XM_017000867.1 | Intron | XP_016856356.1 | ||||
XM_017000868.1 | Intron | XP_016856357.1 |
Set Membership: |
HapMap |