Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGGGCGGCCGTTTAGTCGGCTGA[G/T]GAGAAGCGGACACCAGCGGCGTTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 604844 MIM: 606254 | ||||||||||||||||||||
Literature Links: |
HS2ST1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HS2ST1 - heparan sulfate 2-O-sulfotransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134492.1 | 178 | UTR 5 | NP_001127964.1 | |||
NM_012262.3 | 178 | UTR 5 | NP_036394.1 |
SEP15 - 15 kDa selenoprotein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004261.3 | 178 | Intron | NP_004252.2 | |||
NM_203341.1 | 178 | Intron | NP_976086.1 |