Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCAAGGAAGCCAGAGGTCTGGGTC[A/G]GCCCAAAGCCTCCAAATAGCTCCAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616316 MIM: 611211 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM168A PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FAM168A - family with sequence similarity 168 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286050.1 | Intron | NP_001272979.1 | ||||
NM_001286051.1 | Intron | NP_001272980.1 | ||||
NM_015159.2 | Intron | NP_055974.1 | ||||
XM_017017415.1 | Intron | XP_016872904.1 | ||||
XM_017017416.1 | Intron | XP_016872905.1 |
RELT - RELT tumor necrosis factor receptor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |