Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTTTATTCTGTAGCTAGGGAAAA[A/G]CCATGGAATGGTATCTTAAAAGAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604433 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNE3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
KCNE3 - potassium voltage-gated channel subfamily E regulatory subunit 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005472.4 | Intron | NP_005463.1 | ||||
XM_011544713.2 | Intron | XP_011543015.1 | ||||
XM_017017047.1 | Intron | XP_016872536.1 | ||||
XM_017017048.1 | Intron | XP_016872537.1 | ||||
XM_017017049.1 | Intron | XP_016872538.1 | ||||
XM_017017050.1 | Intron | XP_016872539.1 | ||||
XM_017017051.1 | Intron | XP_016872540.1 | ||||
XM_017017052.1 | Intron | XP_016872541.1 |
Set Membership: |
HapMap |