Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATGCTTGTAATTTATTATTTAGTG[C/T]TTCTTTTCCCTTACAGCAGCCTTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608574 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EMSY PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
EMSY - EMSY, BRCA2 interacting transcriptional repressor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300942.1 | Intron | NP_001287871.1 | ||||
NM_001300943.1 | Intron | NP_001287872.1 | ||||
NM_001300944.1 | Intron | NP_001287873.1 | ||||
NM_020193.4 | Intron | NP_064578.2 | ||||
XM_005274112.4 | Intron | XP_005274169.1 | ||||
XM_006718637.3 | Intron | XP_006718700.1 | ||||
XM_006718638.3 | Intron | XP_006718701.1 | ||||
XM_006718639.3 | Intron | XP_006718702.1 | ||||
XM_011545173.2 | Intron | XP_011543475.1 | ||||
XM_011545174.2 | Intron | XP_011543476.1 | ||||
XM_011545175.2 | Intron | XP_011543477.1 | ||||
XM_011545177.2 | Intron | XP_011543479.1 | ||||
XM_011545178.2 | Intron | XP_011543480.1 | ||||
XM_011545179.2 | Intron | XP_011543481.1 | ||||
XM_017018021.1 | Intron | XP_016873510.1 | ||||
XM_017018022.1 | Intron | XP_016873511.1 |
Set Membership: |
HapMap |