Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTCAAATATGTTTTTCAGGTCCTG[G/T]GTTGCATAGTTTGGATATAGATTCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604080 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF138 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF138 - zinc finger protein 138 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001160183.2 | Intron | NP_001153655.1 | ||||
NM_001271637.1 | Intron | NP_001258566.1 | ||||
NM_001271638.1 | Intron | NP_001258567.1 | ||||
NM_001271639.1 | Intron | NP_001258568.1 | ||||
NM_001271640.1 | Intron | NP_001258569.1 | ||||
NM_001271649.1 | Intron | NP_001258578.1 | ||||
NM_006524.3 | Intron | NP_006515.2 |
Set Membership: |
HapMap |