Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAATTGCTTATATGCCTGGGATCAA[A/G]TGGAGGTCAGGTCTTCTCATCTTGA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
CD200R1L PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CD200R1L - CD200 receptor 1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008784.2 | Intron | NP_001008784.2 | ||||
NM_001199215.1 | Intron | NP_001186144.1 | ||||
XM_011512763.2 | Intron | XP_011511065.1 | ||||
XM_017006300.1 | Intron | XP_016861789.1 |
LOC100129297 - uncharacterized LOC100129297 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |