Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGACTCTAGGCAAATTACCTAACC[A/T]CCGTGTGTTTCAGAAATATTGACAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TMEM128 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TMEM128 - transmembrane protein 128 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297551.1 | Intron | NP_001284480.1 | ||||
NM_001297552.1 | Intron | NP_001284481.1 | ||||
NM_032927.3 | Intron | NP_116316.1 | ||||
XM_005248034.2 | Intron | XP_005248091.1 |