Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601232 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PIK3CG PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PIK3CG - phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit gamma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282426.1 | 120 | Intron | NP_001269355.1 | |||
NM_001282427.1 | 120 | Intron | NP_001269356.1 | |||
NM_002649.3 | 120 | UTR 5 | NP_002640.2 | |||
XM_005250443.3 | 120 | UTR 5 | XP_005250500.1 | |||
XM_011516316.1 | 120 | Intron | XP_011514618.1 | |||
XM_011516317.1 | 120 | Intron | XP_011514619.1 | |||
XM_017012328.1 | 120 | Intron | XP_016867817.1 |
Set Membership: |
HapMap |