Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAATCTGGCACGCAGGTTAAGGAT[C/G]ACCATTCCTAAGGCAAGCAGGTGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602334 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EMP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
EMP2 - epithelial membrane protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001424.5 | 1366 | UTR 3 | NP_001415.1 | |||
XM_006720864.3 | 1366 | UTR 3 | XP_006720927.1 |
Set Membership: |
HapMap |