Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAGGGCCAAGGACAAGGTGCACC[A/G]GTCTAGCGGTCCTCCCGGACCGGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613357 MIM: 609795 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FIBCD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FIBCD1 - fibrinogen C domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145106.1 | 1496 | UTR 3 | NP_001138578.1 | |||
NM_032843.4 | 1496 | UTR 3 | NP_116232.3 |
QRFP - pyroglutamylated RFamide peptide | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |