Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAAGGGATAAATAGAATTTGAGG[A/G]ATTGGTGAGAACGGGTCTATGGATA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 186711 MIM: 607081 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD27 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CD27 - CD27 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242.4 | 195 | Intron | NP_001233.1 | |||
XM_011521042.2 | 195 | UTR 5 | XP_011519344.1 | |||
XM_017020232.1 | 195 | Intron | XP_016875721.1 | |||
XM_017020233.1 | 195 | UTR 5 | XP_016875722.1 | |||
XM_017020234.1 | 195 | Intron | XP_016875723.1 |
CD27-AS1 - CD27 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TAPBPL - TAP binding protein like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |