Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATACTCCGTGCAATACAAAGAACTC[C/T]CAGATGCATCCTGTGGTTTGAGAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176257 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNA6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KCNA6 - potassium voltage-gated channel subfamily A member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002235.3 | 1126 | Intron | NP_002226.1 | |||
XM_005253686.3 | 1126 | UTR 5 | XP_005253743.1 | |||
XM_011520955.2 | 1126 | UTR 5 | XP_011519257.1 | |||
XM_017019270.1 | 1126 | UTR 5 | XP_016874759.1 | |||
XM_017019271.1 | 1126 | UTR 5 | XP_016874760.1 | |||
XM_017019272.1 | 1126 | UTR 5 | XP_016874761.1 |
Set Membership: |
HapMap |