Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGTTGTGAGCTGAGCTTACTACCT[C/T]TTTTTCATAGAACATTCTTTTTACT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616082 MIM: 603070 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C12orf4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
C12orf4 - chromosome 12 open reading frame 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RAD51AP1 - RAD51 associated protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130862.1 | Intron | NP_001124334.1 | ||||
NM_006479.4 | Intron | NP_006470.1 |