Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATCACTGATATATGAGAGTGATA[C/T]GGTAGGAAGAAAACTCAAGCTTATA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
26 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605481 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASPM PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ASPM - abnormal spindle microtubule assembly | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZBTB41 - zinc finger and BTB domain containing 41 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_194314.2 | 6608 | UTR 3 | NP_919290.2 | |||
XM_011509507.2 | 6608 | Intron | XP_011507809.1 | |||
XM_017001205.1 | 6608 | Intron | XP_016856694.1 |
Set Membership: |
HapMap |