Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCAGGTTAACTGGTTATTTTCTT[C/T]TAAGACCTCTTATGAATGAGACTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604642 | ||||||||||||||||||||
Literature Links: |
CA10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CA10 - carbonic anhydrase 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001082533.1 | 2297 | UTR 3 | NP_001076002.1 | |||
NM_001082534.1 | 2297 | UTR 3 | NP_001076003.1 | |||
NM_020178.4 | 2297 | UTR 3 | NP_064563.1 | |||
XM_017024878.1 | 2297 | UTR 3 | XP_016880367.1 |