Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACATTAAACAGGTACAGGTGCTAA[A/G]AAAGTTTAAGACTGATCTTTTGGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604478 | ||||||||||||||||||||
Literature Links: |
CBX5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CBX5 - chromobox 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127321.1 | 8336 | UTR 3 | NP_001120793.1 | |||
NM_001127322.1 | 8336 | UTR 3 | NP_001120794.1 | |||
NM_012117.2 | 8336 | UTR 3 | NP_036249.1 |
MIR3198-2 - microRNA 3198-2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |