Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAACTGATGGCGTTTCATGGTGAA[A/G]CTGTGTTGGATCATCCCAGAGAAGC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609666 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TPCN1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TPCN1 - two pore segment channel 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143819.2 | Intron | NP_001137291.1 | ||||
NM_001301214.1 | Intron | NP_001288143.1 | ||||
NM_017901.5 | Intron | NP_060371.2 | ||||
XM_005253906.4 | Intron | XP_005253963.1 | ||||
XM_011538490.2 | Intron | XP_011536792.1 | ||||
XM_011538492.2 | Intron | XP_011536794.1 | ||||
XM_011538493.2 | Intron | XP_011536795.1 | ||||
XM_017019480.1 | Intron | XP_016874969.1 | ||||
XM_017019481.1 | Intron | XP_016874970.1 | ||||
XM_017019482.1 | Intron | XP_016874971.1 |
Set Membership: |
HapMap |