Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTGAAGTTTCTGATACATGCATT[G/T]ATGTAATACTGGTATTGAAGGCAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609368 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ATL2 - atlastin GTPase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308076.1 | 3177 | UTR 3 | NP_001295005.1 | |||
XM_011533017.1 | 3177 | UTR 3 | XP_011531319.1 | |||
XM_011533018.2 | 3177 | UTR 3 | XP_011531320.1 | |||
XM_011533019.2 | 3177 | UTR 3 | XP_011531321.1 | |||
XM_011533020.1 | 3177 | UTR 3 | XP_011531322.1 | |||
XM_011533022.2 | 3177 | UTR 3 | XP_011531324.1 | |||
XM_011533023.2 | 3177 | UTR 3 | XP_011531325.1 | |||
XM_017004686.1 | 3177 | UTR 3 | XP_016860175.1 | |||
XM_017004687.1 | 3177 | UTR 3 | XP_016860176.1 | |||
XM_017004688.1 | 3177 | UTR 3 | XP_016860177.1 |
Set Membership: |
HapMap |