Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACCACAAACAGGTGAGGAATGCT[A/G]TGACATTACCATGGCTATAATGTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603763 MIM: 602881 | ||||||||||||||||||||
Literature Links: |
KIFC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KIFC1 - kinesin family member C1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002263.3 | Intron | NP_002254.2 | ||||
XM_011514585.1 | Intron | XP_011512887.1 | ||||
XM_011514587.2 | Intron | XP_011512889.1 | ||||
XM_017010836.1 | Intron | XP_016866325.1 | ||||
XM_017010837.1 | Intron | XP_016866326.1 |
PHF1 - PHD finger protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |