Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGGTTGGAGTAATGTACTTGAAT[A/G]TTATTGCCAAATATAATACATATCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RBM46 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RBM46 - RNA binding motif protein 46 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001277171.1 | Intron | NP_001264100.1 | ||||
NM_001277173.1 | Intron | NP_001264102.1 | ||||
NM_144979.4 | Intron | NP_659416.1 | ||||
XM_006714119.2 | Intron | XP_006714182.1 | ||||
XM_006714120.1 | Intron | XP_006714183.1 | ||||
XM_011531692.1 | Intron | XP_011529994.1 | ||||
XM_011531693.1 | Intron | XP_011529995.1 | ||||
XM_011531694.1 | Intron | XP_011529996.1 | ||||
XM_011531695.2 | Intron | XP_011529997.1 | ||||
XM_011531696.1 | Intron | XP_011529998.1 | ||||
XM_011531697.2 | Intron | XP_011529999.1 |
Set Membership: |
HapMap |