Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCAATAAAAGCTTCTATATACTT[A/T]TTCTCCTGTTTTGTACTTGTCATGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606261 MIM: 613940 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NUDT6 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NUDT6 - nudix hydrolase 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPATA5 - spermatogenesis associated 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317799.1 | Intron | NP_001304728.1 | ||||
NM_145207.2 | Intron | NP_660208.2 | ||||
XM_005262783.4 | Intron | XP_005262840.1 | ||||
XM_011531678.2 | Intron | XP_011529980.1 | ||||
XM_011531679.2 | Intron | XP_011529981.1 | ||||
XM_017007825.1 | Intron | XP_016863314.1 | ||||
XM_017007826.1 | Intron | XP_016863315.1 | ||||
XM_017007827.1 | Intron | XP_016863316.1 | ||||
XM_017007828.1 | Intron | XP_016863317.1 | ||||
XM_017007829.1 | Intron | XP_016863318.1 | ||||
XM_017007830.1 | Intron | XP_016863319.1 |
Set Membership: |
HapMap |