Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTCAGCTGTCTTGGTATAAATAC[A/T]CAGCACATGCATTAGTTTGAACCGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300189 MIM: 300311 | ||||||||||||||||||||
Literature Links: |
DLG3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DLG3 - discs large MAGUK scaffold protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TEX11 - testis expressed 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003811.1 | Intron | NP_001003811.1 | ||||
NM_031276.2 | Intron | NP_112566.2 | ||||
XM_011530994.1 | Intron | XP_011529296.1 | ||||
XM_011530996.1 | Intron | XP_011529298.1 | ||||
XM_017029649.1 | Intron | XP_016885138.1 | ||||
XM_017029650.1 | Intron | XP_016885139.1 | ||||
XM_017029651.1 | Intron | XP_016885140.1 | ||||
XM_017029652.1 | Intron | XP_016885141.1 |