Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCAGGGAGCCAGATGAGGCAGAT[A/C]CCATTACCTCTTCTTTTATAAAGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 309845 | ||||||||||||||||||||
Literature Links: |
MSN PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MSN - moesin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002444.2 | Intron | NP_002435.1 | ||||
XM_005262269.2 | Intron | XP_005262326.1 | ||||
XM_011530959.1 | Intron | XP_011529261.1 | ||||
XM_017029545.1 | Intron | XP_016885034.1 | ||||
XM_017029546.1 | Intron | XP_016885035.1 |