Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACAATCCTGATCTCTGACCCATCT[C/T]GAGCAGGAAAGTTGCAAAGCTTCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
16 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608458 MIM: 614428 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NCDN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NCDN - neurochondrin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TFAP2E - transcription factor AP-2 epsilon | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178548.3 | 538 | Intron | NP_848643.2 | |||
XM_017001139.1 | 538 | UTR 5 | XP_016856628.1 |