Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGAGGAGGAATCAAGCTGGTGCCT[A/G]AAATCCCTCCCAGCCCAAAGAATTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
36 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614549 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SERINC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SERINC2 - serine incorporator 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199037.1 | 846 | Intron | NP_001185966.1 | |||
NM_001199038.1 | 846 | Intron | NP_001185967.1 | |||
NM_001199039.1 | 846 | UTR 5 | NP_001185968.1 | |||
NM_018565.3 | 846 | Intron | NP_061035.2 | |||
NM_178865.4 | 846 | Intron | NP_849196.2 |