Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTGTTCATGTATCTGCTGCATAT[A/T]AAATAATGAATGGGTTTTAATTTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603125 | ||||||||||||||||||||
Literature Links: |
TPST1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TPST1 - tyrosylprotein sulfotransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003596.3 | Intron | NP_003587.1 | ||||
XM_005250642.2 | Intron | XP_005250699.1 | ||||
XM_011516634.2 | Intron | XP_011514936.1 | ||||
XM_011516635.2 | Intron | XP_011514937.1 | ||||
XM_017012724.1 | Intron | XP_016868213.1 | ||||
XM_017012725.1 | Intron | XP_016868214.1 | ||||
XM_017012726.1 | Intron | XP_016868215.1 | ||||
XM_017012727.1 | Intron | XP_016868216.1 |