Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGCTTCCCCGCCTCCTTCTGGAA[C/T]AGCGCGTACCAGGCGCCAGTGCCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609979 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
VGLL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
VGLL2 - vestigial like family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153453.1 | 464 | Intron | NP_703154.1 | |||
NM_182645.3 | 464 | Silent Mutation | AAC,AAT | N,N 147 | NP_872586.1 | |
XM_005266883.2 | 464 | Silent Mutation | AAC,AAT | N,N 147 | XP_005266940.1 |
Set Membership: |
HapMap |