Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607898 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRIB3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TRIB3 - tribbles pseudokinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301188.1 | Intron | NP_001288117.1 | ||||
NM_001301190.1 | Intron | NP_001288119.1 | ||||
NM_001301193.1 | Intron | NP_001288122.1 | ||||
NM_001301196.1 | Intron | NP_001288125.1 | ||||
NM_001301201.1 | Intron | NP_001288130.1 | ||||
NM_021158.4 | Intron | NP_066981.2 | ||||
XM_017027989.1 | Intron | XP_016883478.1 |
Set Membership: |
HapMap |