Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 182390 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SCN2A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SCN2A - sodium voltage-gated channel alpha subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040142.1 | Intron | NP_001035232.1 | ||||
NM_001040143.1 | Intron | NP_001035233.1 | ||||
NM_021007.2 | Intron | NP_066287.2 | ||||
XM_005246753.3 | Intron | XP_005246810.1 | ||||
XM_005246755.4 | Intron | XP_005246812.1 | ||||
XM_017004655.1 | Intron | XP_016860144.1 | ||||
XM_017004656.1 | Intron | XP_016860145.1 | ||||
XM_017004657.1 | Intron | XP_016860146.1 | ||||
XM_017004658.1 | Intron | XP_016860147.1 | ||||
XM_017004659.1 | Intron | XP_016860148.1 |
Set Membership: |
HapMap |