Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTACCTTTTTATCCTTCCACCCTA[A/G]GCTTCTCTCCTCCCTCTTCCCTCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 182340 MIM: 607321 | ||||||||||||||||||||
Literature Links: |
ATP1A2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ATP1A2 - ATPase Na+/K+ transporting subunit alpha 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ATP1A4 - ATPase Na+/K+ transporting subunit alpha 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001734.1 | 203 | Intron | NP_001001734.1 | |||
NM_144699.3 | 203 | UTR 5 | NP_653300.2 | |||
XM_011509582.1 | 203 | Intron | XP_011507884.1 |