Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGACGTGAGTACTTAACTCTAGAGG[G/T]TTGTAAGAATTAAATAGTGTCTGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600938 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RBBP6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
RBBP6 - RB binding protein 6, ubiquitin ligase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006910.4 | Intron | NP_008841.2 | ||||
NM_018703.3 | Intron | NP_061173.1 | ||||
NM_032626.5 | Intron | NP_116015.2 |
Set Membership: |
HapMap |