Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTATTATGTACTACACGCCGGGA[C/G]TACCTCAAGAGGTGAAACAAACAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF546 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF546 - zinc finger protein 546 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297763.1 | 1189 | Intron | NP_001284692.1 | |||
NM_178544.4 | 1189 | Intron | NP_848639.2 | |||
XM_011526898.2 | 1189 | Missense Mutation | CTA,GTA | L,V 7 | XP_011525200.1 | |
XM_011526899.2 | 1189 | Intron | XP_011525201.1 |