Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGAGTCTGCAGTGTGCCTAACGCG[A/T]ACTAGAGGGACCGAGGCGGGGCGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605576 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SMC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SMC2 - structural maintenance of chromosomes 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042550.1 | Intron | NP_001036015.1 | ||||
NM_001042551.1 | Intron | NP_001036016.1 | ||||
NM_001265602.1 | Intron | NP_001252531.1 | ||||
NM_006444.2 | Intron | NP_006435.2 | ||||
XM_006716933.3 | Intron | XP_006716996.1 | ||||
XM_011518148.2 | Intron | XP_011516450.1 | ||||
XM_011518149.2 | Intron | XP_011516451.1 | ||||
XM_011518150.2 | Intron | XP_011516452.1 | ||||
XM_011518151.2 | Intron | XP_011516453.1 | ||||
XM_011518152.2 | Intron | XP_011516454.1 | ||||
XM_011518153.1 | Intron | XP_011516455.1 | ||||
XM_017014206.1 | Intron | XP_016869695.1 | ||||
XM_017014207.1 | Intron | XP_016869696.1 | ||||
XM_017014208.1 | Intron | XP_016869697.1 | ||||
XM_017014209.1 | Intron | XP_016869698.1 | ||||
XM_017014210.1 | Intron | XP_016869699.1 | ||||
XM_017014211.1 | Intron | XP_016869700.1 | ||||
XM_017014212.1 | Intron | XP_016869701.1 |
SMC2-AS1 - SMC2 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |