Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607527 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTBP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PTBP3 - polypyrimidine tract binding protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163788.2 | 5319 | UTR 3 | NP_001157260.1 | |||
NM_001163790.2 | 5319 | UTR 3 | NP_001157262.1 | |||
NM_001244896.1 | 5319 | UTR 3 | NP_001231825.1 | |||
NM_001244897.1 | 5319 | Missense Mutation | ACA,GCA | T,A 545 | NP_001231826.1 | |
NM_001244898.1 | 5319 | UTR 3 | NP_001231827.1 | |||
NM_005156.6 | 5319 | UTR 3 | NP_005147.3 | |||
XM_006717346.1 | 5319 | UTR 3 | XP_006717409.1 | |||
XM_011519265.1 | 5319 | UTR 3 | XP_011517567.1 | |||
XM_017015365.1 | 5319 | UTR 3 | XP_016870854.1 |