Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCTTTTACCTTGATTGGTGACAT[C/T]CTCCCATGCTTCCAGTGGACTGGTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612869 | ||||||||||||||||||||
Literature Links: |
ATRNL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ATRNL1 - attractin like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001276282.3 | Intron | NP_001263211.1 | ||||
NM_207303.4 | Intron | NP_997186.1 | ||||
XM_011539587.1 | Intron | XP_011537889.1 | ||||
XM_011539588.2 | Intron | XP_011537890.1 | ||||
XM_011539589.2 | Intron | XP_011537891.1 | ||||
XM_011539590.2 | Intron | XP_011537892.1 | ||||
XM_011539591.2 | Intron | XP_011537893.1 | ||||
XM_017016035.1 | Intron | XP_016871524.1 | ||||
XM_017016036.1 | Intron | XP_016871525.1 | ||||
XM_017016037.1 | Intron | XP_016871526.1 | ||||
XM_017016038.1 | Intron | XP_016871527.1 | ||||
XM_017016039.1 | Intron | XP_016871528.1 | ||||
XM_017016040.1 | Intron | XP_016871529.1 |