Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCACTTTTAAATGAGCATTTCAGA[C/T]TGCTTGTTTAGCCATGAAAAGTCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603946 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HELLS PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HELLS - helicase, lymphoid-specific | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289067.1 | Intron | NP_001275996.1 | ||||
NM_001289068.1 | Intron | NP_001275997.1 | ||||
NM_001289069.1 | Intron | NP_001275998.1 | ||||
NM_001289070.1 | Intron | NP_001275999.1 | ||||
NM_001289071.1 | Intron | NP_001276000.1 | ||||
NM_001289072.1 | Intron | NP_001276001.1 | ||||
NM_001289073.1 | Intron | NP_001276002.1 | ||||
NM_001289074.1 | Intron | NP_001276003.1 | ||||
NM_001289075.1 | Intron | NP_001276004.1 | ||||
NM_018063.4 | Intron | NP_060533.2 |