Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATTCCTGGTCCAGTACTCTAGCG[C/T]GGCCTGGATGGAGATGGGTGGTGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114070 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANXA6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ANXA6 - annexin A6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001155.4 | 2721 | UTR 3 | NP_001146.2 | |||
NM_001193544.1 | 2721 | UTR 3 | NP_001180473.1 | |||
XM_005268432.4 | 2721 | UTR 3 | XP_005268489.1 |
Set Membership: |
HapMap |