Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGACTATTAACTTTCACTCTTTA[C/T]ACTCTTTAATACTTGGATGTCAGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607888 MIM: 609906 MIM: 605658 MIM: 160710 | ||||||||||||||||||||
Literature Links: |
CMTM5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CMTM5 - CKLF like MARVEL transmembrane domain containing 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
EFS - embryonal Fyn-associated substrate | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IL25 - interleukin 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022789.3 | Intron | NP_073626.1 | ||||
NM_172314.1 | Intron | NP_758525.1 |
MYH6 - myosin heavy chain 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |