Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTTTTAATGCATGTTGAAGGCAT[C/T]TAAGTAGTGTTTCCAGAGTGCATTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603113 MIM: 608973 | ||||||||||||||||||||
Literature Links: |
PPP2R1B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PPP2R1B - protein phosphatase 2 scaffold subunit Abeta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001177562.1 | Intron | NP_001171033.1 | ||||
NM_001177563.1 | Intron | NP_001171034.1 | ||||
NM_002716.4 | Intron | NP_002707.3 | ||||
NM_181699.2 | Intron | NP_859050.1 | ||||
NM_181700.1 | Intron | NP_859051.1 | ||||
XM_017017958.1 | Intron | XP_016873447.1 | ||||
XM_017017959.1 | Intron | XP_016873448.1 | ||||
XM_017017960.1 | Intron | XP_016873449.1 | ||||
XM_017017961.1 | Intron | XP_016873450.1 |
SIK2 - salt inducible kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015191.2 | Intron | NP_056006.1 | ||||
XM_017017417.1 | Intron | XP_016872906.1 | ||||
XM_017017418.1 | Intron | XP_016872907.1 |