Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGATAAGTAACCATTGTGTAAAAT[A/G]TTTCTGCTAACTTAATGTGCAGTAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
OR8B2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
OR8B2 - olfactory receptor family 8 subfamily B member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005468.1 | Intron | NP_001005468.1 | ||||
XM_017017535.1 | Intron | XP_016873024.1 | ||||
XM_017017536.1 | Intron | XP_016873025.1 |
OR8B3 - olfactory receptor family 8 subfamily B member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005467.1 | Intron | NP_001005467.1 | ||||
XM_017017716.1 | Intron | XP_016873205.1 | ||||
XM_017017717.1 | Intron | XP_016873206.1 | ||||
XM_017017718.1 | Intron | XP_016873207.1 |
Set Membership: |
HapMap |