Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGATCTGTGTTTGAATCCTGGCTCT[A/G]CCACTAGTTTTTGCAACTTGGGCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
OR8B3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
OR8B3 - olfactory receptor family 8 subfamily B member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005467.1 | Intron | NP_001005467.1 | ||||
XM_017017716.1 | Intron | XP_016873205.1 | ||||
XM_017017717.1 | Intron | XP_016873206.1 | ||||
XM_017017718.1 | Intron | XP_016873207.1 |
Set Membership: |
HapMap |