Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGACCTGAGAGATGATGGACAGAGA[A/G]GTGACGTGGAAAAAGCCACCAACCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613066 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PKNOX2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PKNOX2 - PBX/knotted 1 homeobox 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022062.2 | Intron | NP_071345.2 | ||||
XM_005271642.2 | Intron | XP_005271699.1 | ||||
XM_005271643.2 | Intron | XP_005271700.1 | ||||
XM_006718894.2 | Intron | XP_006718957.1 | ||||
XM_011542944.2 | Intron | XP_011541246.1 | ||||
XM_011542945.2 | Intron | XP_011541247.1 | ||||
XM_011542946.1 | Intron | XP_011541248.1 | ||||
XM_011542947.2 | Intron | XP_011541249.1 | ||||
XM_017018110.1 | Intron | XP_016873599.1 | ||||
XM_017018111.1 | Intron | XP_016873600.1 | ||||
XM_017018112.1 | Intron | XP_016873601.1 |
PKNOX2-AS1 - PKNOX2 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |