Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACCAATGTCAATAGGGTGGCTGA[C/T]GGGGCCTAGATTTGCTACCAGATAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 114130 | ||||||||||||||||||||
Literature Links: |
CALCA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CALCA - calcitonin related polypeptide alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033952.2 | 1951 | Intron | NP_001029124.1 | |||
NM_001033953.2 | 1951 | UTR 3 | NP_001029125.1 | |||
NM_001741.2 | 1951 | Intron | NP_001732.1 | |||
XM_017018283.1 | 1951 | UTR 3 | XP_016873772.1 | |||
XM_017018284.1 | 1951 | UTR 3 | XP_016873773.1 |