Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGGGTCCAGGACCCCCCTTCAGA[G/T]GGCTGAGGAGCAGTTGAGGGTGAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EVI5L PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
EVI5L - ecotropic viral integration site 5 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159944.2 | Intron | NP_001153416.1 | ||||
NM_145245.4 | Intron | NP_660288.1 | ||||
XM_005272458.4 | Intron | XP_005272515.1 | ||||
XM_017026239.1 | Intron | XP_016881728.1 |
Set Membership: |
HapMap |