Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTCAGACCGCGTGGCCCCCGTGG[A/G]CGGTGCGTGGCATGGGGGTGGGCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608258 | ||||||||||||||||||||
Literature Links: |
DPP9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DPP9 - dipeptidyl peptidase 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_139159.4 | 464 | Intron | NP_631898.3 | |||
XM_005259673.2 | 464 | Intron | XP_005259730.1 | |||
XM_011528405.1 | 464 | Intron | XP_011526707.1 | |||
XM_011528406.1 | 464 | Intron | XP_011526708.1 | |||
XM_011528408.1 | 464 | Intron | XP_011526710.1 | |||
XM_011528410.1 | 464 | Intron | XP_011526712.1 | |||
XM_017027453.1 | 464 | Intron | XP_016882942.1 | |||
XM_017027454.1 | 464 | Intron | XP_016882943.1 |
DPP9-AS1 - DPP9 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242901.1 | 464 | Missense Mutation | ACG,GCG | T,A 50 | NP_001229830.1 |